Order Entry
United States
ContactUsLinkComponent
DRG® PTH (Parathyroid) Intact ELISA, DRG International
DRG® PTH (Parathyroid) Intact ELISA, DRG International
Catalog # 75871-400
Supplier:  DRG International
CAS Number:  
undefined
DRG® PTH (Parathyroid) Intact ELISA, DRG International
Catalog # 75871-400
Supplier:  DRG International
Supplier Number:  EIA-3645
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Host:
  • Primary antibody reactivity:
  • Description:
    PTH (Parathyroid) Intact ELISA
  • Environmentally Preferable:
  • Detection Range:
    0.9 pg/ml
  • Application:
    Endocrinology
  • Regulatory Status:
    FDA Regulated; CE Approved
  • Cat. No.:
    75871-400
  • Supplier no.:
    EIA-3645
  • No. of tests:
    96 wells

Specifications

About this item

An enzyme immunoassay for the quantitative determination of Intact-PTH (Parathyroid Hormone) in serum.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

PTH (Parathyroid hormone, Parathormone, Parathyrin) is biosynthesized in the parathyroid gland as a pre-proparathyroid hormone, a larger molecular precursor consisting of 115 amino acids. Following sequential intracellular cleavage of a 25-amino acid sequence, preproparathyroid hormone is converted to an intermediate, a 90-amino acid polypetide, proparathyroid hormone. With additional proteolytic modification, proparathyroid hormone is then converted to parathyroid hormone, an 84 amino acid polypeptide. In healthy individuals, regulation of parathyroid hormone secretion normally occurs via a negative feedback action of serum calcium on the parathyroid glands. Intact PTH is biologically active and clears very rapidly from the circulation with a half-life of less than four minutes1. PTH undergoes proteolysis in the parathyroid glands, but mostly peripherally, particularly in the liver but also in the kidneys and bone, to give N-terminal fragments and longer lived C‑terminal and midregion fragments. In subjects with renal insufficiency, C‑terminal and midregion PTH assays typically give elevated PTH results, as reflected by impaired renal clearance2. The DRG Intact PTH Immunoassay is a two-site ELISA [Enzyme-Linked ImmunoSorbent Assay] for the measurement of the biologically intact 84 amino acid chain of PTH. Two different goat polyclonal antibodies to human PTH have been purified by affinity chromatography to be specific for well-defined regions on the PTH molecule. One antibody is prepared to bind only the mid-region and C-terminal PTH 39-84 and this antibody is biotinylated. The other antibody is prepared to bind only the N-terminal PTH 1-34 and this antibody is labeled with horseradish peroxidase [HRP] for detection. Although mid-region and C-terminal fragments are bound by the biotinylated anti-PTH (39-84), only the intact PTH 1‑84 forms the sandwich complex necessary for detection. The capacity of the biotinylated antibody and the streptavidin coated microwell both have been adjusted to exhibit negligible interference by inactive fragments, even at very elevated levels. In this assay, calibrators, controls, or patient samples are simultaneously incubated with the enzyme labeled antibody and a biotin coupled antibody in a streptavidin-coated microplate well. At the end of the assay incubation, the microwell is washed to remove unbound components and the enzyme bound to the solid phase is incubated with the substrate, tetramethylbenzidine (TMB). An acidic stopping solution is then added to stop the reaction and converts the color to yellow. The intensity of the yellow color is directly proportional to the concentration ofintact PTH in the sample. A dose response curve of absorbance unit vs. concentration is generated using results obtained from the calibrators. Concentrations of intact PTH present in the controls and patient samples are determined directly from this curve.