Order Entry
United States
ContactUsLinkComponent
DRG® Treponema pallidum IgG ELISA, DRG International
DRG® Treponema pallidum IgG ELISA, DRG International
Catalog # 75871-196
Supplier:  DRG International
CAS Number:  
undefined
DRG® Treponema pallidum IgG ELISA, DRG International
Catalog # 75871-196
Supplier:  DRG International
Supplier Number:  EIA-3517R
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    Treponema pallidum IgG ELISA
  • Environmentally Preferable:
  • Application:
    Infectious Diseases
  • Regulatory Status:
    FDA Regulated; CE Approved
  • Cat. No.:
    75871-196
  • Supplier no.:
    EIA-3517
  • No. of tests:
    96 wells

Specifications

About this item

An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Treponema pallidum in serum.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

Spirochetes are motile bacteria with a periplasmatic axial filament

Three further stages of disease follow which are classified as secondary, tertiary, and quaternary syphilis. Treatment with antibiotics at the earliest disease stage and prophylactic measures are ways to prevent epidemics. For this purpose, antenatal and donor blood screenings are mandatory in most of countries around the world.The DRG Treponema pallidum IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Treponema pallidum antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Treponema pallidum-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgG antibodies are dispensed into the wells. During a second incubation this anti‑IgG conjugate binds specifically to IgG antibodies resulting in the formation of enzyme-linked immune complexes.After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Treponema pallidum-specific IgG antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.

All pathogenic species belong to the family Treponemataceae, which includes the three genera: Treponema, Borrelia, and Leptospira. The Treponema are motile bacteria, 5-15µ in length and 0.2µ in width, containing about 10 flexible, undulating, spiral shaped rods. Treponema pallidum, the causative agent of Syphilis, is transmitted by direct contact, usually through sexual intercourse. Syphilis along with Gonorrhoea, Chancroid and Lymphogranuloma venereum, designated as a venereal disease, or VD, is an acute and chronic infectious disease. After an incubation period of 12-30 days, the first symptoms to appear are chancres, soon followed by syphilitic ulcers which then spontaneously disappear in a few weeks. During this first stage (primary syphilis) the Treponema pallidum propagates in related lymph nodes to be distributed to the whole body stream.