Order Entry
United States
ContactUsLinkComponent
DRG® Candida albicans IgA ELISA, DRG International, Inc.
DRG® Candida albicans IgA ELISA, DRG International, Inc.
Catalog # 75871-260
Supplier:  DRG International
CAS Number:  
undefined
DRG® Candida albicans IgA ELISA, DRG International, Inc.
Catalog # 75871-260
Supplier:  DRG International
Supplier Number:  EIA-3457R
CAS Number:  

Specifications

  • Description:
    Candida albicans IgA ELISA
  • Sample Type:
    Serum
  • Application:
    Infectious Diseases
  • Regulatory Status:
    FDA Regulated; CE Approved
  • Cat. No.:
    75871-260
  • Supplier no.:
    EIA-3457
  • No. of tests:
    96 wells

Specifications

About this item

An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Candida albicans in serum.

  • High quality assays with reproducible and reliable results
  • Ready-to-use reagents with internal controls
  • Very good precision and sensitivity
  • Short assay time and incubation steps at room temperature
  • Simple and technician friendly tests

The DRG Candida albicans IgA ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with Candida albicans antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation Candida albicans-specific antibodies of positive specimens and controls are bound to the immobilized antigens. After a washing step to remove unbound sample and control material horseradish peroxidase conjugated anti-human IgA antibodies are dispensed into the wells. During a second incubation this anti‑IgA conjugate binds specifically to IgA antibodies resulting in the formation of enzyme-linked immune complexes. After a second washing step to remove unbound conjugate the immune complexes formed (in case of positive results) are detected by incubation with TMB substrate and development of a blue color. The blue color turns into yellow by stopping the enzymatic indicator reaction with sulfuric acid. The intensity of this color is directly proportional to the amount of Candida albicans-specific IgA antibody in the patient specimen. Absorbance at 450 nm is read using an ELISA microtiter plate reader.