To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Assay Type:Sandwich
- Host:
- Primary antibody reactivity:
- Description:Prolactin ELISA
- Environmentally Preferable:
- Sample Type:Serum
- Detection Range:0.35 – 200 ng/ml
- Application:Fertility/Pregnancy
- Regulatory Status:Not FDA Regulated; CE Approved
- Cat. No.:75871-254
- Supplier no.:EIA-1291
- No. of tests:96 wells
Specifications
About this item
An enzyme immunoassay for the quantitative measurement of prolactin in serum.
- High quality assays with reproducible and reliable results
- Ready-to-use reagents with internal controls
- Very good precision and sensitivity
- Short assay time and incubation steps at room temperature
- Simple and technician friendly tests
Human prolactin (lactogenic hormone) is secreted from the anterior pituitary gland in both men and women
The DRG Prolactin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site on a Prolactin molecule. An aliquot of sample containing endogenous Prolactin is incubated in the coated well with enzyme conjugate, which is an anti- Prolactin antibody conjugated with horseradish peroxidase. After incubation the unbound conjugate is washed off. The amount of bound peroxidase is proportional to the concentration of Prolactin in the sample. Having added the substrate solution, the intensity of color developed is proportional to the concentration of Prolactin in the sample.
Human prolactin is a single chain polypeptide hormone with a molecular weight of approximately 23.000 daltons. The release and synthesis of prolactin is under neuroendocrinal control, primarily through Prolactin Releasing Factor and Prolactin Inhibiting Factor. Women normally have slightly higher basal prolactin levels than men; apparently, there is an estrogen- related rise at puberty and a corresponding decrease at menopause. The primary functions of prolactin are to initiate breast development and to maintain lactation. Prolactin also suppresses gonadal function. During pregnancy, prolactin levels increase progressively to between 10 and 20 times normal values, declining to non-pregnant levels by 3-4 weeks post- partum. Breast feeding mothers maintain high levels of prolactin, and it may take several months for serum concentrations to return to non-pregnant levels. The determination of prolactin concentration is helpful in diagnosing hypothalamic- pituitary disorders. Microadenomas (small pituitary tumors) may cause hyperprolactinemia, which is sometimes associated with male impotence. High prolactin levels are commonly associated with galactorrhea and amenorrhea. Prolactin concentrations have been shown to be increased by estrogens, thyrotropin- releasing hormone (TRH), and several drugs affecting dopaminergic mechanisms. Prolactin levels are elevated in renal disease and hypothyroidism, and in some situations of stress, exercise, and hypoglycemia. Additionally, the release of prolactin is episodic and demonstrates diurnal variation. Mildly elevated prolactin concentrations should be evaluated taking these considerations into account. Prolactin concentrations may also be increased by drugs such as chloropromazine and reserpine, and may be lowered by bromocyptine and L-dopa.The DRG Prolactin ELISA provides a rapid, sensitive, and a reliable assay. The antibodies developed for the test will determine a minimal concentration of human prolactin of 0.35 ng/mL. There is no cross-reactivity with hCG, TSH, LH, FSH, or hGH.