Order Entry
United States
ContactUsLinkComponent
 
Tissue-Tek® AutoSection® Automated Microtome, Sakura Finetek
  10204-886
undefined
Tissue-Tek® AutoSection® Automated Microtome, Sakura Finetek
  10204-886
 :  5010
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    Auto-Section® Automated Microtome
  • Environmentally Preferable:
  • Electrical:
    115VAC, 50/60 Hz, Single phase, 7.1 amps
  • W×D×H:
    15×26.4×17.4"
  • Weight:
    121 lbs.
  • Cat. No.:
    10204-886
  • Supplier no.:
    5010

 

 

AutoSection automatically aligns block to the blade edge, saving tissue and technician time.

  • Auto-aligns block to blade edge, even if block was previously sectioned on a different microtome
  • One touch trimming and sectioning elimate repetitive hand wheel motions and reduces the risk of musculoskeletal disorders
  • 3D chuck/specimen holder
  • LCD touch screen control panel
  • Wireless remote control
  • One-touch operation to engage cutting modes
  • 16 user-designed sectioning programs

Tissue-Tek® AutoSection® is the most precise microtome on the market as determine by lasar measurements

Multiple built-in safeguards make AutoSection® the leader in microtome user safety. One-touch trimming and sectioning eliminate repetitive handwheel motions and reduces the risk of musculoskeletal disorders.

Programmable sectioning enables laboratories to implement standardized protocols for specific tissue or block types. AutoSection® also standardizes section thickness to help optimise advanced staining procedures such as IHC and ISH. Patented AutoAlign and AutoTrim features assure preservation of tissue as well as saving technician's time.