Jump to:
About this item
For the chromogenic detection and enumeration of Enterococci in water by the membrane filter technique
Conforms with US EPA Approved Methods
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
For the chromogenic detection and enumeration of Enterococci in water by the membrane filter technique
Conforms with US EPA Approved Methods
Documents
Product Family Options
Product Information
- SizeAvailabilityPrice
- 90000-790m EI Agar, BD DifcoSupplier Number: 214881Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Discontinued-Supplier Number: -