SafTest Dispenser Sets, MP Biomedicals, LLC
Supplier: MP Biomedicals
Certificates
About this item
Pre-set volumetric dispensers which allow for quick analysis.
- Choose the dispenser set for each chosen endpoint
- Allows for accurate testing, every time
- For use with SafTest kits
Peroxide Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, C, and Preparation Reagent) and one reagent holding rack.
Free Fatty Acid Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Malonaldehyde Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Alkenal Test Kit Dispenser Set includes one dispenser for each Reagent (A, B, and Preparation Reagent) and one reagent holding rack.
Total Glycerin Test Kit Dispenser Set includes one dispenser for each Reagent (A and Preparation Reagent) and one reagent holding rack.
Caution: Storage: Room temperature
Frequently Bought Together