SafTest Test Kits, MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
MP Biomedicals' Test Kits are designed to measure alkenal, free fatty acid, malonaldehyde, percent fat, peroxide or total glycerin content in any liquid or solid sample containing oil/fat.
- Chose from six different kits
- For use with SafTest instrument and consumable platforms
- Results in as little as 30 minutes
MP Biomedicals' Test Kits contains all reagents and calibrators necessary to run samples. A control is also included in each Test Kit. Each kit will allow for testing of approximately 100 samples. Sample dilution will likely be required for appropriate analysis.
Specifications
- Detection Method:Bichromatic photometer
- Storage Temperature:2 - 8 °C
Specifications
Related Information
Frequently Bought Together