Order Entry
United States
ContactUsLinkComponent
Luciferase SPG/T7 Control DNA, Promega
Luciferase SPG/T7 Control DNA, Promega
Catalog # PAL4821
CAS Number:  
undefined
Luciferase SPG/T7 Control DNA, Promega
Catalog # PAL4821
Supplier Number:  L4821
CAS Number:  

Specifications

  • Description:
    Luciferase T7 Control DNA, 20microg
  • Size:
    20 μg
  • Storage Conditions:
    Store at –20°C
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAL4821
  • Supplier no.:
    L4821

Specifications

About this item

Luciferase SP6 and T7 Control DNAs are used with the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter.

  • Functional Controls for the TnT(R) Quick Coupled and TnT(R) Coupled Transcription/Translation Systems
  • Contains luciferase gene under transcriptional control of an SP6 or T7 phage RNA polymerase promoter
  • Includes a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene
  • Easy to monitor control reactions with a luciferase assay

The Luciferase SP6 and T7 Control DNAs are used as functional controls in the TNT Quick Coupled and TNT Coupled Transcription/Translation Systems. The Control DNAs contain the gene for luciferase under transcriptional control of a phage RNA polymerase promoter. All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are monitored easily by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components. Luciferase Control DNAs are supplied as 0.5mg/ml solutions in TE buffer.

Luciferase Control DNAs are supplied as 0.5 mg/mL solutions in TE buffer.

All constructs carry a 30-base pair poly[d(A)/d(T)] tail following the luciferase gene. Control reactions are easily monitored by the production of luminescence, which is generated from full-length luciferase and the addition of necessary components.