Order Entry
United States
ContactUsLinkComponent
 
Thermolysin, Promega
Catalog # PAV4001
Supplier:  Avantor
CAS Number:  
Thermolysin, Promega
Catalog # PAV4001
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    25 mg
  • Storage Conditions:
    –30...–10 °C
  • Protein/Peptide Name:
    Thermolysin
  • Cat. No.:
    PAV4001
  • Supplier no.:
    V4001

Specifications

About this item

Thermolysin is a thermostable metalloproteinase. The high digestion temperatures may be used as an alternative to denaturants to improve digestion of proteolytically resistant proteins. The optimal digestion temperature range is 65-85°C.

  • Thermostable Proteinase with High Digest Temperature; Better Denaturation, Digestion of Proteolytically Resistant Proteins
  • Cleaves at the N-terminus of Leu, Phe, Val, Ile, Ala, Met
  • Optimal digestion temperature is 65-85°C
  • Optimal activity at pH 5.0-8.5

Thermolysin is a thermostable metalloproteinase. The high digestion temperatures may be used as an alternative to denaturants to improve digestion of proteolytically resistant proteins. Thermolysin preferentially cleaves at the N-terminus of the hydrophobic residues leucine, phenylalanine, valine, isoleucine, alanine and methionine. The optimal digestion temperature range is 65-85°C. Thermolysin activity is optimal at pH 5.0-8.5.

The optimal digestion temperature range is 65–85°C. Thermolysin activity is optimal at pH 5.0–8.5.