Order Entry
United States
Orders LinkContactUsLinkComponent
BBL™ LBS Agar, BD Life Sciences

BBL™ LBS Agar, BD Life Sciences

LBS Agar, 500 g
Catalog # 90000-152
Supplier:   BD
Supplier #:  211327
BBL™ LBS Agar, BD Life Sciences
Catalog # 90000-152
Supplier:  BD
CAS Number:  
BBL™ LBS Agar, BD Life Sciences

LBS Agar, 500 g

CAS Number:  
Price
$300.96
New Customer Promotional Price
Log in for availability
Each (1lb)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Selective isolation and enumeration of lactobacilli.

Specifications

Specifications

  • Description:
    LBS Agar
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.