To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:SSC Buffer, 20X, Molecular Grade
- Size:1000 mL
- Environmentally Preferable:
- Storage Temperature:15 to 30 °C
- Formulation:3 M NaCl, 0.3 M sodium citrate (for 20X concentration); lead: ≤10 ppm
- Cat. No.:PAV4261
- pH:6.9 - 7.1 (25 °C, 20X)
- Supplier no.:V4261
Specifications
About this item
SSC Buffer, 20X, Molecular Grade is commonly used in nucleic acid hybridization techniques at concentrations from 0.1X to 20X, depending on the application.
- Quality tested and certified free of DNase, RNase and protease activity
It has a clear, colorless liquid form.
Conductivity at 25°C (2X): 24.4 – 32.4 mmhos"