Order Entry
United States
ContactUsLinkComponent
QuantiFluor RNA System, 1ml, Promega
QuantiFluor RNA System, 1ml, Promega
Catalog # PAE3310
CAS Number:  
undefined
QuantiFluor RNA System, 1ml, Promega
Catalog # PAE3310
Supplier Number:  E3310
CAS Number:  

Specifications

  • Description:
    QuantiFluor RNA System, 1ml
  • Size:
    1ml
  • Storage Conditions:
    Store at –30°C to –10°C, protected from light.
  • Storage Temperature:
    -30°C to -10°C
  • Cat. No.:
    PAE3310
  • Supplier no.:
    E3310

Specifications

About this item

The QuantiFluor RNA System contains a fluorescent RNA-binding dye that enables sensitive quantitation of small amounts of RNA in solution.

  • Sensitive Quantitation of Small Amounts of RNA in Solution
  • Requires less template RNA than other systems
  • More sensitive than absorbance methods for low-concentration samples

Sensitive quantitation of RNA is important for the success of downstream applications. The QuantiFluor RNA System contains a fluorescent RNA-binding dye that enables sensitive quantitation of small amounts of RNA in solution. Detecting and quantitating small amounts of RNA is a very important step that is used in many biological applications, particularly in molecular biology techniques.

Detecting and quantitating small amounts of RNA is an important step used in many biological applications particularly molecular biology techniques.