ContactUsLinkComponent
GloSensor cAMP Reagent, Promega
GloSensor cAMP Reagent, Promega
Certificates
undefined
GloSensor cAMP Reagent, Promega
Certificates
Specifications
  • Storage Conditions:
    Store the GloSensor cAMP Reagent at –70°C, and store the pGloSensor cAMP Plasmid at –20°C. Store the re–suspended reagent at –70°C in single–use aliquots.
  • Storage Temperature:
    LESS THAN -65°C