Order Entry
United States
ContactUsLinkComponent
 
Maxwell® RSC SimplyRNA Kits, Promega
Catalog # PAAS1380
CAS Number:  
undefined
Maxwell® RSC SimplyRNA Kits, Promega
Catalog # PAAS1380
Supplier Number:  AS1380
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Magnetic beads
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Blood (dried) on filter cards
    Blood
    Biofluids (serum, plasma, saliva)
    Cultured cells
    FFPE tissues
    Human/animal cells and tissues
  • Target molecule:
    RNA
  • Description:
    Maxwell RSC simplyRNA Blood Kit, 48 preps
  • Size:
    48 preps
  • Environmentally Preferable:
  • Storage Conditions:
    Upon receipt, remove 1–Thioglycerol and store at 2 to 10°C. Store the remaining kit components at room temperature (15 to 30°C).
  • Description:
    Maxwell RSC simplyRNA Blood Kit
  • Cat. No.:
    PAAS1380
  • Supplier no.:
    AS1380

Specifications

About this item

The Maxwell RSC simplyRNA Kits, used with the Maxwell RSC Instrument, provides a simple method for automated RNA purification from a variety of sample types. 

  • For RNA Extraction from Blood, Tissue, and Cells to Use in Gene Expression and Translation
  • Consistent Results and Sufficient Yield for Downstream Assays
  • Versatile Protocol
  • All Reagents and Consumables, Including DNase and Plastics, Provided
  • Simple Processing Steps

The low elution volume is used to generate concentrated high-quality RNA suitable for use in downstream applications such as quantitative RT-PCR

The Maxwell RSC Instrument can process from one to 16 samples in under an hour.

The kit provides the necessary reagents for processing the samples and uses prefilled cartridges for purification to maximize simplicity and convenience.

Accessories information: Maxwell 16 LEV Cartridge Rack Available Separately, One Each