About this item
Unparalleled control of volumetric and serological pipettes from 0.1 to 200 ml.
- Smooth control with pressure-sensitive buttons and thumbwheel speed limiter
- Quiet, lightweight, balanced and extremely low vibration
- 8-hour full-speed operation on a single charge
- Indicator lights when 2 hours of charge remaining
- Gravity/powered dispensing switch
- Autoclavable 0.2 µm sterile filter
Lightweight controllers weigh only 190 g (6.6 oz.) and have slim handgrip for comfort during serial pipetting, even for small hands.
Easily replaceable NiMH battery provides eight full hours of continuous pipetting on a single four-hour charge. An LED charge indicator alerts user when two hours are left.
Pressure-sensitive buttons provide smooth control over pipetting speed. An additional control adjusts maximum speed for both accurate pipetting of small volumes and speedy movement of large volumes. Dual-mode switch provides either powered dispensing, gravity dispensing for volumetric work, or density gradient underlaying. Pipette adapter and 0.2 µm filter are autoclavable for sterile use.