About this item
The AMP-Glo Assay is a homogeneous assay that generates a luminescent signal from any biochemical reaction that produces AMP as a reaction product.
- Monitor AMP Concentration in Biochemical Reactions
- Use with virtually with any AMP-producing enzyme
- Well-suited for low-activity enzymes
- Sensitive to low AMP concentration
The AMP-Glo Assay is a homogeneous assay that generates a luminescent signal from any biochemical reaction that produces AMP as a reaction product. This versatile system can measure the activity of a broad range of enzymes, such as cyclic AMP-specific phosphodiesterases, aminoacyl-tRNA synthetases, DNA ligases and ubiquitin ligases or enzymes modulated by AMP. The AMP-Glo Assay is designed to quantitatively monitor the concentration of AMP in a biochemical reaction in a wide range of plate formats, including high-throughput formats. The stable luminescent signal of the assay eliminates the need for an injector-equipped luminometer and allows batch-mode processing of multiple plates. The assay can be used to determine the AMP produced either in the presence or absence of ATP as a substrate. The assay contains two reagents: one to terminate the AMP-generating enzymatic reaction and simultaneously remove ATP and convert AMP produced into ADP, and a second reagent that converts the ADP to ATP followed by conversion of the ATP into a luminescent signal using the luciferin/luciferase reaction. The assay also is well suited for monitoring AMP produced in biochemical reactions catalyzed by enzymes that do not use ATP as a substrate, such as cAMP-dependent phosphodiesterases (PDE) and bacterial DNA ligases. The AMP-Glo Assay has a high dynamic range and produces a strong signal at low substrate conversion, making it well suited for screening low activity enzymes. The assay produces minimal false hits and Z´ values greater than 0.7.
Specifications
- Storage Conditions:Store the system at –30 to –10 °C. Before using the components, thaw all of them completely at room temperature except for the AMP–Glo Reagent II, which should be kept on ice after thawing. Once thawed, mix all components thoroughly before use. Once prepared, the Kinase–Glo One Solution should be dispensed into aliquots and stored at –20 °C. See the product label for expiration date.
- Storage Temperature:-30°C to -10°C