Order Entry
United States
ContactUsLinkComponent
GSH/GSSG-Glo Assay, 10 mL, Promega
GSH/GSSG-Glo Assay, 10 mL, Promega
Certificates
undefined
GSH/GSSG-Glo Assay, 10 mL, Promega
Certificates

Specifications

  • Storage Conditions:
    Store at –20 °C and away from light.
  • Storage Temperature:
    -30°C to -10°C

Specifications

Related Information

Frequently Bought Together