Order Entry
United States
ContactUsLinkComponent
 
Arg-C
  PAV1881
 :  
undefined
Arg-C
  PAV1881
 :  V1881
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Size:
    10 μg
  • Storage Conditions:
    Store at 2...10 °C
  • Enzyme Name:
    Arg-C
  • Grade:
    Sequencing grade
  • Cat. No.:
    PAV1881
  • Supplier no.:
    V1881

 

 

Arg-C (clostripain) is an endopeptidase that cleaves at the C-terminus of arginine residues, including the sites next to proline. Cleavage also will occur at lysine residues. Arg-C activity is optimal in the pH range of 7.6-7.9.

  • Sequencing-Grade Protease: for use Alone or with other Proteases for Mass Spectrometry Analysis
  • Cleaves at the C-terminus of arginine residues, including sites next to proline
  • Also cleaves at lysine residues
  • Activity is optimal in the pH range of 7.6-7.9

Arg-C (clostripain) is an endopeptidase that cleaves at the C-terminus of arginine residues, including the sites next to proline. Cleavage also will occur at lysine residues. This sequencing grade enzyme can be used alone or in combination with other proteases for protein analysis by mass spectrometry and other applications. Arg-C activity is optimal in the pH range of 7.6-7.9.

Arg-C activity is optimal in the pH range of 7.6–7.9.

This sequencing-grade enzyme can be used alone or in combination with other proteases for protein analysis by mass spectrometry and other applications.