Two-Stop Peristaltic Pump Tubing for PerkinElmer ICP Instruments, PerkinElmer
Supplier: PerkinElmer
Certificates
About this item
PerkinElmer offers a wide range of high-quality peristaltic pump tubing for all your AA, ICP-OES and ICP-MS applications.
- Compatible with PerkinElmer Optima® and Avio® ICP-OES Instruments and NexION® ICP-MS Instruments
- Pump tubing should be inspected and replaced on a regular basis as part of a preventative maintenance schedule
- Maintain factory performance using genuine PerkinElmer replacement parts
Remember to replace your tubing frequently and always keep spares on hand.
Frequently Bought Together