Maxwell® 16 Viral Total Nucleic Acid Purification Kit, Promega
Catalog # PAAS1150
Supplier: Promega Corporation
CAS Number:
Specifications
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Viral RNA
- Sample type:Biofluids (serum, plasma, saliva)
- Target molecule:RNA
- Description:Maxwell 16 Viral Total Nucleic Acid Purification Kit, 48 preps
- Size:48 preps
- Storage Conditions:Store at 15 to 30°C.
- Description:Maxwell 16 Viral Total Nucleic Acid Purification Kit
- Cat. No.:PAAS1150
- Supplier no.:AS1150
Specifications
About this item
The Maxwell 16 Viral Total Nucleic Acid Purification Kit is designed for automated extraction of viral total nucleic acid (RNA and DNA) from serum, plasma or other samples using the Maxwell 16 Instrument.
- Kit Provides General Purpose Viral Total Nucleic Acid Extraction from Serum, Plasma, and Other Sample Types
- Purified Nucleic Acids May Be Used in Applications Such as qPCR and qRT-PCR
- Single Kit and Protocol for Multiple Sample Types
- No Detectable Cross-Contamination
These sample types are commonly processed in molecular microbiology or virology areas of molecular diagnostics
The simple protocol involves three main steps. First, lysis buffer and proteinase K are mixed to prepare a lysis solution. Second, lysis solution is mixed with sample. Third, the lysate is added into the cartridges. Purified viral total nucleic acids are ready for analysis in approximately 45 minutes.
The kit contains all the necessary reagents in a convenient prefilled cartridge format.
Accessories information: Maxwell 16 LEV Cartridge Rack Available Separately, One Each