Maxwell® 16 FFPE Plus LEV DNA Purification Kit, Promega
PAAS1135
:
- Method / Format:Magnetic beads
- Prep size:Mini
- Refine target molecule:Total DNA
- Sample type:FFPE tissues
- Target molecule:DNA
- Description:Maxwell 16 FFPE Plus LEV DNA Purification Kit, 48 preps
- Size:48 preps
- Storage Conditions:Store at 15 to 30°C.
- Description:Maxwell 16 FFPE Plus LEV DNA Purification Kit
- Cat. No.:PAAS1135
- Supplier no.:AS1135
The Maxwell 16 FFPE Plus LEV DNA Purification Kit is designed to extract DNA rapidly from FFPE samples using the Maxwell 16 Instrument.
- For Automated Genomic DNA Purification
- Sufficient Yield for Downstream Amplification
- Immediate Time Savings
- Overnight and One-Hour Options for Digest
- No Organic Solvents
The FFPE Kit is specifically designed for optimal purification of DNA from one to ten 5 μm-thin sections of formalin-fixed paraffin-embedded (FFPE) tissue samples
Lysates are placed directly into the cartridges, and genomic DNA is ready for amplification in approximately 30 minutes after proteinase K digest. This kit does not use hazardous xylene, and quality testing demonstrates virtually no PCR inhibitors in purified samples.
The kit is optimized to yield a final DNA concentration that maximizes DNA yield, eliminating the need to concentrate the extract prior to amplification.
: Maxwell 16 LEV Cartridge Rack Available Separately, One Each