CellTiter 96 AQueous MTS Reagent Powder, Promega
Supplier: Promega Corporation
Certificates
About this item
CellTiter 96(R) AQueous MTS Reagent Powder is a novel tetrazolium compound for use in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays.
- A Novel Tetrazolium Compound for Use in Colorimetric Assays
- Available in two sizes
- Use in assays to determine the number of viable cells
CellTiter 96(R) AQueous MTS Reagent Powder is a novel tetrazolium compound for use in colorimetric assays to determine the number of viable cells in proliferation, cytotoxicity or chemosensitivity assays. It can be purchased separately and is provided in powdered form.
Specifications
- Storage Temperature:2°C to 10°C