Order Entry
United States
ContactUsLinkComponent
Wizard SV 96 Plasmid DNA Purification System, Promega
Wizard SV 96 Plasmid DNA Purification System, Promega
Certificates
undefined
Wizard SV 96 Plasmid DNA Purification System, Promega
Certificates

Specifications

  • Method / Format:
    Multiwell plate
  • Prep size:
    Mini
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Sample Size:
    1–1.5 mL of culture
  • Description:
    Wizard SV 96 Plasmid DNA Purification System
  • Yield:
    3–5 µg

Specifications

Related Information

Frequently Bought Together