About this item
The ROS-Glo H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions.
- Specific, Direct Detection of Hydrogen Peroxide (H2O2)
- Fast assay involves a simple two-reagent-addition protocol
- Substrate reacts directly with H2O2; no need for HRP as a coupling enzyme
The ROS-Glo H2O2 Assay is a homogeneous, fast and sensitive bioluminescent assay that measures the level of hydrogen peroxide (H2O2), a reactive oxygen species (ROS), directly in cell culture or in defined enzyme reactions. A derivatized luciferin substrate is incubated with sample and reacts directly with H2O2 to generate a luciferin precursor. Addition of ROS-Glo Detection Solution converts the precursor to luciferin and provides Ultra-Glo Recombinant Luciferase to produce light signal that is proportional to the level of H2O2 present in the sample.
The addition of ROS-Glo Detection Solution converts the precursor to luciferin and provides Ultra-Glo™ Recombinant Luciferase to produce light signal that is proportional to the level of H2O2 present in the sample.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Conditions:Store all components at –30°C to –10°C.
- Storage Temperature:-30°C to -10°C