About this item
GoTaq G2 Flexi DNA Polymerase is supplied with 5X Green GoTaq Flexi Buffer and 5X Colorless GoTaq Flexi Buffer and 25mM MgCl2.
- High-Performance GoTaq(R) G2 DNA Polymerase with Mg-Free Buffer System
- Full-length Taq DNA polymerase that exhibits 5´→3´ exonuclease activity
- Direct to gel convenience
- Flexibility to optimize Mg concentration in PCR
- Supplied at a concentration of 5u/microl
The second generation of GoTaq products, GoTaq G2 Flexi DNA Polymerase reliably amplifies a wide range of PCR templates and provides high-performing results due to improved manufacturing processes, increased reliability and consistency. GoTaq G2 Flexi DNA Polymerase is supplied with 5X Green GoTaq Flexi Buffer and 5X Colorless GoTaq Flexi Buffer and 25mM MgCl2. The GoTaq G2 Flexi DNA Polymerases are supplied in a proprietary formulation containing 50% glycerol with buffers designed for enhanced amplification. The enzyme is a full-length form of Taq DNA polymerase that exhibits 5'-->3' exonuclease activity. The 5X Green GoTaq Reaction and Flexi Buffers contain two dyes (blue and yellow) that separate during electrophoresis to indicate migration progress. The colorless buffer is used when direct fluorescence or absorbance readings are required without prior purification of the amplified DNA from the PCR.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: