Order Entry
United States
Orders LinkContactUsLinkComponent
LPM Agar Base, BD™

LPM Agar Base, BD™

Supplier:   BD
LPM Agar Base, BD™
Supplier:  BD
Certificates
LPM Agar Base, BD™

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Use with Moxalactam Antimicrobic supplement for isolation of L

monocytogenes.

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 90001-602
    LPM Agar Base
    LPM Agar Base
    2 kg
    Log in for availability
    Log in for availability
    Each (2kg)
    Each (2kg) - $1,475.10
    $1,475.10
    New Customer Promotional Price
    Each (2kg)
       $1,475.10
    Minimum order quantity is 1

    Supplier Number: 222110
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • LPM Agar Base
    500 g
    Discontinued
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.