Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Supplier: BD
Certificates
About this item
Difco™ Fluid Lactose Medium is used in microbiology for the cultivation of Salmonella and coliform organisms in water, food, dairy and pharmaceutical products.
- Salmonella cultivation
- Coliform organism cultivation
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.