ContactUsLinkComponent
Lab Clamp, Heavy Duty, Glas-Col®, LLC
Lab Clamp, Heavy Duty, Glas-Col®, LLC
Catalog # 33796-002
Supplier:  Glas-Col
Small Business Enterprise
undefined
Lab Clamp, Heavy Duty, Glas-Col®, LLC
Catalog # 33796-002
Supplier:  Glas-Col
Supplier Number:  100DLC300
Small Business Enterprise
Specifications
  • Description:
    Lab Clamp
  • Shipping Weight:
    1 lb.
  • Cat. No.:
    33796-002
  • For use with:
    Rods up to 3/4" dia.
  • Supplier no.:
    100DLC300
Specifications
About this item
Right angle clamp with adjustment knobs.
  • Sturdy construction
  • Easily attached
  • Easy adjustment
This right angle lab clamp is ideal for heavy duty use on 3/4" rods.
Delivery: 7-10 days after order.