Orders LinkContactUsLinkComponent
Quantus™ Fluorometer, Promega
Quantus™ Fluorometer, Promega
Quantus™ Fluorometer, Promega
Quantus™ Fluorometer, Promega
Quantus™ Fluorometer, Promega
The Quantus Fluorometer is a dual-channel fluorometer for your personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate.
  • Simple, sensitive DNA and RNA quantitation
  • Easy, intuitive controls
  • Ready to use with QuantiFluor(R) dyes
  • Recommended for next-generation sequencing
The Quantus Fluorometer is a dual-channel fluorometer for your personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate. The Quantus Fluorometer is optimized with preprogrammed settings for Promega QuantiFluor Dye Systems (QuantiFluor dsDNA, RNA and ssDNA Systems) to quantitate nucleic acids and offers the flexibility to create customized methods and quantitation settings for other fluorescent dyes. The Quantus Fluorometer is equipped with two fluorescence channels for nucleic acid and protein quantitation: 1) Blue fluorescence channel: Excitation 495nm shortpass (wavelengths up to 495nm), emission 510-580nm; 2) Red fluorescence channel: Excitation 640nm shortpass (wavelengths up to 640nm), emission 660-720nm.
Ordering information: To learn more about the Quantus Fluorometer, please contact your Avantor representative.
Services: The Quantus Fluorometer comes with a 1-year warranty upon purchase of the instrument. Return a completed product registration card within 90 days of purchase to receive an additional 1 year of warranty free of charge. An additional 1-year warranty may be purchased (Cat.# SA4040). Warranty coverage includes all parts, labor and shipping to and from our depot repair locations. The warranty does not include repair or replacement necessitated by accident, neglect, misuse, unauthorized repair or modification of the instrument. If a problem arises with the instrument, please contact Promega Technical Services for support. If further action is required, you will be given repair options and assigned a return authorization number. Promega is not responsible for instrumentation returned without an authorization number.
Documents
Product Family Options
Services
  • Description
    Wavelength
  • Quantus Fluorometer 1-Year Service Agreement
    Each
    Each - $784.54
    $784.54
    Each
       $784.54
    Minimum order quantity is 1
     : SA4040
  • Description
    Wavelength
  • Quantus™ Fluorometer
    260 nm
    Each
    Each - $2,598.27
    $2,598.27
    Each
       $2,598.27
    Minimum order quantity is 1
     : E6150
Services
  •  
    Description
  • PASA4040
    Quantus Fluorometer 1-Year Service Agreement
    Each - $784.54