Order Entry
United States
ContactUsLinkComponent
3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98%, solid
3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98%, solid
Catalog # 100266-656
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
3-Methyl-2-benzothiazolinonehydrazone hydrochloride ≥98%, solid
Catalog # 100266-656
Supplier Number:  171411GM
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    100266-656
  • Supplier no.:
    171411GM

Specifications

About this item

 :
 :215.70 g/mol
Melting Pt:275…277 °C
UN:12352100
ADR:,
CAS Number:4338-98-1
Formula:C₈H₁₀ClN₃S
MDL Number:MFCD00005816