Order Entry
United States
ContactUsLinkComponent
4',5,7-Trihydroxy-3'-methoxyflavanone ≥99% (by HPLC)
4',5,7-Trihydroxy-3'-methoxyflavanone ≥99% (by HPLC)
Catalog # 100263-494
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
4',5,7-Trihydroxy-3'-methoxyflavanone ≥99% (by HPLC)
Catalog # 100263-494
Supplier Number:  021118S10MG
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 mg
  • Cat. No.:
    100263-494
  • Supplier no.:
    021118S10MG

Specifications

About this item

 :
 :302.29 g/mol
Melting Pt:225…227 °C
UN:12352100
ADR:,
CAS Number:107657-60-3
Formula:C₁₆H₁₄O₆
MDL Number:MFCD00017314