ContactUsLinkComponent
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Catalog # 101975-897
Supplier:  Oxyrase Inc
Brucella Pea And KVL OxyPRAS Plus Multi-Pack, Oxyrase, Inc.
Catalog # 101975-897
Supplier:  Oxyrase Inc
Supplier Number:  M-BKP-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    Pack of 10
  • Environmentally Preferable:
  • Cat. No.:
    101975-897
  • Storage temperature:
    +2…+8 °C
  • Supplier no.:
    M-BKP-3
Specifications
About this item
Individually packaged BRU, PEA and KVL plates; banded together. OxyPRAS® Plates are specifically made to improve recovery of anaerobes.
  • Specifically made for anaerobes to improve recovery of injured cells
  • Arrive reduced and ready to use
  • Stays reduced in air for hours
  • Saves tech time and helps eliminate errors
  • Specially packaged in oxygen barrier bags
  • Store refrigerated for a extended shelf life or at room temperature
  • Meets Best Practice criteria for working with clinical anaerobes
Due to the Oxyrase® Enzyme System, OxyPRAS Plus® Plates (Pre-Reduced Anaerobically Sterilized) are technologically advanced compared to other plates.
Oxyrase® utilized during the manufacture of the plate prevents oxidation during its preparation. Oxyrase® is again added to the final plate. Oxyrase® keeps the plate reduced and confers unique properties to the OxyPRAS Plus® plate not found in standard or conventional PRAS plates. PRAS plates are considered the gold standard to isolate all anaerobes from clinical specimens.
OxyPRAS Plus® Multi-Packs are individually wrapped plates; banded together. OxyPRAS Plus® Multi-Packs save time, help eliminate errors, add value and flexibility by being banded together. Each plate has its own lot number. Six month shelf-life when refrigerated and three month expiration date at room temperature.