ContactUsLinkComponent
HemoCue Glucose 201 Data Management System, HemoCue America
HemoCue Glucose 201 Data Management System, HemoCue America
Catalog # 10842-846
Supplier:  Hemocue
undefined
HemoCue Glucose 201 Data Management System, HemoCue America
Catalog # 10842-846
Supplier:  Hemocue
Supplier Number:  121422
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    HemoCue Glucose (GL) 201 DM Analyzer
  • Cat. No.:
    10842-846
  • Supplier no.:
    121422
Specifications
About this item
The GL201 DM System offers lab accurate POC glucose testing with all the necessary connectivity solutions to seamlessly transfer results into your electronic database.
  • CLIA - Waived
  • Small Sample Size - 5 uL
  • Measurement Range 0 - 444 mg/dL
  • Lab Accurate
  • Capillary or Venous Blood
  • Barcode Scanner
  • QC Lockout
  • Bi-Directional Communication
With its unique cuvette technology, the HemoCue® Glucose 201 DM system gives you high accuracy at the Point-of-Care
The system safeguards Patient Testing and Data with customizable automatic prompts for patient ID, operator ID, lot numbers, etc. User log-in and lockout functions, quality control tests including QC lockout, linearity, and proficiency testing.
Used for diagnosis and monitoring of diabetes mellitus. Microcuvette technology means no need to bring analyzer near patients, reducing the risk of spreading infection. Individually wrapped microcuvettes to avoid contamination and maximize shelf life (vial cuvettes are also available to high-throughput users).
Plus, you have the added controls and productivity means made possible with advanced data management. With instant data connection, customizable prompts, and more, you gain tighter control of authorized use and higher efficiency in everything from patient management to billing.
Certifications: CLIA Waived.
Ordering information: Analyzer, docking stations, software, and cuvettes all sold separately.
Glucose 201 DM Cuvettes
Product Information