About this item
The Hb201 DM System offers lab accurate POC hemoglobin testing with all the necessary connectivity solutions to seamlessly transfer results into your electronic database.
- CLIA - Waived
- Small Sample Size - 10 uL
- Measurement Range 0 - 25.6 g/dL
- Lab Accurate
- Results in under 1 minute
- Barcode Scanner
- QC Lockout
- Bi-Directional Communication
HemoCue’s Hb 201 DM system offers the lab accuracy and ease of use that has become a standard in Hb point-of-care testing, with features like QC lockout and built-in barcode scanning for patient ID to support patient safety and privacy
Seamlessly interface with existing network using POCT-1A, facilitate billing, and help ensure regulatory compliance.
Functions include Operator ID, Patient ID, Lot ID, Site ID, Patient comments, Critical value alerts, STAT testing.
Through bi-directional communication and flexible settings, the Hb 201 DM system offers a complete data management solution for Hb testing and anemia screening in hospitals, primary care and public health.