About this item
This VECTASTAIN® Elite® ABC system is the most sensitive avidin/biotin-based peroxidase system and is particularly important in the localization of antigens present in low amounts or in cases where the cost of the primary antibodies is significant.
- Smaller, very uniform, and highly active, allowing more accessibility for binding to a biotinylated target
- Higher sensitivity leads to lower cost per slide
- Used to detect any molecule that is biotinylated
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. The ABC kit system is modular and different variations can accommodate a wide array of primary antibody and tissue species. This kit uses horseradish peroxidase (HRP) in a compact but highly active ABC detection complex. Several kits also use a secondary antibody to target primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation.