- Description:Western Blue Stabilized Substrate for Alkaline Phosphatase, 100ml
- Size:100ml
- Storage Conditions:Store at 22 to 25 °C
- Storage Temperature:15°C to 30°C
- Cat. No.:PAS3841
- Supplier no.:S3841
Stable, ready-to-use substrate for Western blots and immunoscreening.
- Ready-to-use formulation that does not require dilution or reagent mixing
- As sensitive as other commercially available BCIP/NBT formulations and reagents
- Stable for one year at room temperature
Western Blue Stabilized Substrate for Alkaline Phosphatase is a stable, ready-to-use substrate for Western blots and immunoscreening. It is a mixture of 5-bromo-4-chloro-3-indolyl-phosphate (BCIP) and nitro blue tetrazolium (NBT) in a proprietary stabilizing buffer. Western Blue Substrate should be used directly and without dilution. This liquid substrate deposits a permanent dark purple stain on membrane sites bearing alkaline phosphatase. Western Blue Substrate is as sensitive as other reagents based on the BCIP/NBT formulation.
This liquid substrate deposits a permanent dark purple stain on membrane sites bearing alkaline phosphatase. Western Blue Substrate is as sensitive as other reagents based on the BCIP/NBT formulation.
Western Blue Substrate should be used directly and without dilution.