Justrite® 5-Gallon Portable Mixing Tank
10861-066
: Justrite
- Description:Mixing Tank, 5 Gallon, Red
- Height:16"
- Cat. No.:10861-066
- Depth:15
- Width:15"
- Supplier no.:27705
Fully removable cover makes this container ideal for mixing operations.
- Swinging bail handle makes tank easy to carry
- Ideal for mixing operations
- Fully removable cover