Order Entry
United States
ContactUsLinkComponent
Anti-IgG Goat Antibody
Anti-IgG Goat Antibody
Catalog # 103254-838
Supplier:  ADVANSTA INC MS
Anti-IgG Goat Antibody
Catalog # 103254-838
Supplier:  ADVANSTA INC MS
Supplier Number:  R-05057-250

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody Type:
    Secondary
  • Antigen Name:
    Immunoglobulin G
  • Antigen Symbol:
    IgG
  • Host:
    Goat
  • Reactivity:
    Chicken
  • Size:
    250 µl
  • Cat. No.:
    103254-838
  • Supplier no.:
    R-05057-250

Specifications

About this item