Order Entry
United States
ContactUsLinkComponent
 
180 QT Portable Analyzer, Pointe Scientific
  10023-514
180 QT Portable Analyzer, Pointe Scientific
  10023-514
 :  90-P4504-QT

 

  • Communication Interface:
    USB mouse, USB 2.0 compatible
  • Description:
    Microprocessor controlled analyzer
  • Electrical:
    115 V or 230 V, 50 – 60 Hz
  • W×D×H:
    9" (24 cm)×13.5" ( 34 cm)×5" (13 cm)
  • Sample Volume:
    1 mL
  • Weight:
    10 lbs.
  • Cat. No.:
    10023-514
  • Contents:
    Touch screen interface, 37 °C on-board heating block with 12 wells and a built-in printer
  • Supplier no.:
    90-P4504-QT

 

 

A compact, microprocessor controlled analyzer that can read and calculate results of endpoint, kinetic, initial rate and colorimetric assays. Ideal for teaching future laboratory professionals the fundamentals of clinical chemistry testing

  • Portable
  • Lightweight
  • Light Source: Tungsten lamp with lamp-saver feature
  • Standard Wavelengths: 340, 405, 505, 545, 580 and 630 nm
  • Display: Interactive touch screen 3.5” LCD color graphic display
  • Printer: Thermal dot matrix with graphic capability

Microprocessor used are eZ80, 64K EEPROM. It Stores all parameters including wavelengths, calculations, unit codes, linear and normal ranges, rate timing, standard values, test names, and previous standard curve.

 : For research and education use only.