ContactUsLinkComponent
m-Endo Media Ampoules, Advantec
m-Endo Media Ampoules, Advantec
  101105-196
 :  
m-Endo Media Ampoules, Advantec
  101105-196
 :  COLI200
 :  
  • Pack type:
    50 ampoules/box
  • Description:
    Media Ampoules m-Endo
  • Cat. No.:
    101105-196
  • Volume:
    50×2 ml
  • Supplier no.:
    COLI200
m-Endo Media Ampules are used with membrane filters to culture bacteria, yeasts, and molds
  • Single use vials assure sterility
  • Culture bacteria, yeasts and molds
  • Certified to comply with established test criteria
Available ready-to-use 2 ml ampoules contain enough medium to saturate one standard absorbent pad in a petri dish. Single use vials assure sterility. Each product is lot tested and certified to comply with established test criteria.
 : Quality certificates are included with each shipment, and packages are clearly marked with expiration date.