Phosphate Buffered Saline (PBS) 20X, Genetex
Catalog # 89261-496
Supplier: Genetex
CAS Number:
Specifications
- Description:Phosphate Buffered Saline, 20X
- Size:500 mL
- Cat. No.:89261-496
- Supplier no.:GTX48888
Specifications
About this item
This solution is sufficient to prepare 10 L of 1X phosphate buffered saline
Tested applications: ELISA, FACS, ICC/IF, IHC, WB.
At 25 °C, 1X PBS will have a pH between 7.4 and 7.6. Not intended for any diagnostic or therapeutic purpose. Not suitable for human or animal consumption. Store at room temperature.