Orders LinkContactUsLinkComponent
Zombie NIR™ Fixable Viability Kit, BioLegend
Zombie NIR™ Fixable Viability Kit, BioLegend
Supplier:   Biolegend
Zombie NIR™ Fixable Viability Kit, BioLegend
Supplier:  Biolegend
Certificates
Zombie NIR™ Fixable Viability Kit, BioLegend
Zombie NIR™ Fixable Viability Kit, BioLegend
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Zombie NIR™ Fixable Viability kit is used in flow cytometric analysis.
  • Composed of lyophilized Zombie NIR™ dye and anhydrous DMSO
  • Analysis of intracellularly-stained cells and antibody surface-stained cells
Zombie NIR™ Fixable Viability kit is composed of lyophilized Zombie NIR™ dye and anhydrous DMSO and is used in flow cytometric analysis of intracellularly-stained cells and antibody surface-stained cells.
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 10761-492
    Fixable Viability Kit
    Fixable Viability Kit
    100 Tests
    Log in for availability
    Log in for availability
    Each
    Each - $128.00
    $128.00
    Each
       $128.00
    Minimum order quantity is 1
    Supplier Number: 423105
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 10761-294
    Fixable Viability Kit
    Fixable Viability Kit
    500 Tests
    Log in for availability
    Log in for availability
    Each
    Each - $479.00
    $479.00
    Each
       $479.00
    Minimum order quantity is 1
    Supplier Number: 423106
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.