ContactUsLinkComponent
Gardner Color Standard #4, Aqua Solutions
  103194-014
 :  
undefined
Gardner Color Standard #4, Aqua Solutions
  103194-014
 :  3914-500ML
 :  
  • Pack type:
    Acid glass bottle, amber
  • Description:
    Gardner Color Std. #4
  • Size:
    500 ml
  • Cat. No.:
    103194-014
  • Supplier no.:
    3914-500ML
The solution contains potassium platinum chloride, hydrochloric acid 36-38% and water.