Order Entry
United States
ContactUsLinkComponent
Propidium iodide solution 50 µg/ml in PBS pH 7.4, Calbiochem®
Propidium iodide solution 50 µg/ml in PBS pH 7.4, Calbiochem®
Catalog # 80510-636
Supplier:  MilliporeSigma
CAS Number:  
Propidium iodide solution 50 µg/ml in PBS pH 7.4, Calbiochem®
Catalog # 80510-636
Supplier:  MilliporeSigma
Supplier Number:  537060-5ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic ampoule
  • Size:
    5 ml
  • Cat. No.:
    80510-636
  • Supplier no.:
    537060-5ML

Specifications

Specification Test Results

  • Formulation:
    50 µg/ml in PBS, pH 7.4
  • Primary Target:
    Membrane impermeable DNA intercalator
  • Sample Type:
    Intact Cells
  • Ship Code:
    Ambient Temperature Only
  • Toxicity:
    Standard Handling
  • Storage:
    +2°C to +8°C
  • Protection:
    Protect from Light
  • Do not freeze:
    Ok to freeze
  • Special Instructions:
    Following initial use, aliquot and freeze (−20 °C)

Specification Test Results

About this item

 :
 :668.4 g/mol
Storage Temperature:Refrigerator
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921