Order Entry
United States
ContactUsLinkComponent
 
Ethanol 70%, meets analytical specification of USP (140 Proof)
Catalog # 76212-358
Supplier:  Decon Labs
CAS Number:  
Ethanol 70%, meets analytical specification of USP (140 Proof)
Catalog # 76212-358
Supplier:  Decon Labs
Supplier Number:  2432
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    12×32 oz.
  • Environmentally Preferable:
  • Cat. No.:
    76212-358

Specifications

Specification Test Results

  • Assay-Ethyl Alcohol (%):
    68.0 - 72.0
  • Specific Gravity @ 15.56:
    0.8848 - 0.8948
  • Water (%):
    28.0 - 32.0
  • Color:
    Passes USP Test for Color
  • Clarity:
    Passes USP Test for Clarity
  • Acidity:
    30
  • Alkalinity:
    Passes USP Test
  • Non-volatile Residue:
    =2.5
  • Volatile Impurities (Acetaldehyde):
    =10
  • Volatile Impurities (Benzene):
    =2
  • Volatile Impurities (Methanol):
    =200
  • Volatile Impurities (Other Impurities):
    =300
  • UV Absorbance Curve at 235 - 340 nm run against Water:
    Passes USP for Smooth Curve
  • UV Absorbance at 240 nm:
    =0.40
  • UV Absorbance at 250 nm:
    =0.30
  • UV Absorbance at 260 nm:
    =0.30
  • UV Absorbance at 270 - 340 nm:
    =0.10

Specification Test Results

About this item

Synonyms: Ethyl alcohol
 :
 :46.07 g/mol
UN:1170
 :GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568
Boiling Pt:78.3
Melting Pt:-117
Flash Pt:12
ADR:3 III
Merck Index:12,03806