About this item
Formulated products are avaliable in 91% and 60% USP IPA with USP Water for Injection (WFI), as well as an aseptically filtered 99% USP IPA, ideal for pharmaceutical, biotechnology, healthcare, and medical device cleaning rotation for use in both aseptic and non-aseptic environments.
- Prefiltered to 0.2 um
- Completely traceable and validated for sterility and shelf life
- 60% and 91% formulated with USP Water for Injection, and are gamma irradiated; 99% manufactured via aseptic filtration.
99% delivered with a Certificate of Analysis and Certificate of Sterility.