BT Lab Systems Dry Bath/Heat Block Incubator, G-Biosciences
A microprocessor-controlled dry bath incubator
- LCD display
- Programmable feature to allow automatic control of heating and timing
- Low energy consumption and no noise
- Built in temperature calibration function, automatic fault detection and buzzer alarm function
- Built in over-temperature protection device, safe and reliable use, prolong the service life of the machine
- Product design, compact and tight, small occupied space, freer and easier
- Multiple optional heating blocks are available (See Related Accessories)
A dry bath incubator with precise temperature control, making it a convenient replacement for traditional water baths.
Choice of dry bath incubator or dry bath incubator with both heating and cooling features.
: Accessory Heat blocks are not supplied with the equipment and must be ordered separately.
: No accessory items are supplied with this equipment and must be ordered separately.
- Catalog No:
- 10856-448
- 10856-446
- Heating Time:
- <15 min. for 20-100°C
- Temperature Uniformity:
- 0.3°C
- Timer:
- 99 h, 59 min.
Frequently Bought Together