Order Entry
United States
ContactUsLinkComponent
Tissue Culture Disk Head for Mini Rotator, Glas-Col
Tissue Culture Disk Head for Mini Rotator, Glas-Col
Catalog # 33725-046
Supplier:  Glas-Col
Small Business Enterprise
undefined
Tissue Culture Disk Head for Mini Rotator, Glas-Col
Catalog # 33725-046
Supplier:  Glas-Col
Supplier Number:  099ATC132
Small Business Enterprise

Specifications

  • Description:
    Tissue Culture Disk Head with 30 spring clamps for 5-9mm and 8-14 mm tubes, for Mini Rotator
  • Environmentally Preferable:
  • Cat. No.:
    33725-046
  • Shipping weight:
    2 lbs.
  • Supplier no.:
    099ATC132

Specifications

About this item

Aluminum disk with 30 position clips for 8 mm to 14 mm tubes.

  • Designed for quick change
  • Large and small micro centrifuge and micro test tube applications

Interchangeable disk for use with the mini rotator which quickly changes to increase your capabilities.

Ordering information: Tissue culture disk head with spring clamps.