ContactUsLinkComponent
BT Lab Systems Digital Magnetic Stirrer with Timer, G-Biosciences
BT Lab Systems Digital Magnetic Stirrer with Timer, G-Biosciences
Catalog # 10858-500
Supplier:  G-Biosciences
BT Lab Systems Digital Magnetic Stirrer with Timer, G-Biosciences
Catalog # 10858-500
Supplier:  G-Biosciences
Supplier Number:  BT1005
Specifications
  • Description:
    Digital Magnetic Stirrer with Timer
  • Cat. No.:
    10858-500
  • Supplier no.:
    BT1005
Specifications
About this item
A digital magnetic stirrer designed for stirring <3L containers at a maximum speed of 1,800rpm.
  • DC brushless motor for stable operation, low noise, long life and no risk of sparks
  • Stainless steel shell, firm and durable, thermo stability and anti-corrosion
  • Separate digital display for speed and timer
Digital timer included.